missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Avidin Protein
A cDNA sequence encoding the Avidin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10525-25mg
Additional Details : Weight : 0.01000kg
Specifications
396260 | |
25 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
16 units/mg protein, 1 unit binds 1microg biotin. | |
Other | |
Untagged | |
Greater than 92.0%. |
Avidin Protein | |
Research Use Only | |
AVD | |
Recombinant Protein | |
Non-species Specific | |
MVHATSPLLL LLLLSLALVA PGLSARKCSL TGKWTNDLGS NMTIGAVNSR GEFTGTYITA VTATSNEIKE SPLHGTQNTI NKRTQPTFGF TVNWKFSEST TVFTGQCFID RNGKEVLKTM WLLRSSVNDI GDDWKATRVG INIFTRLRTQ KE |