missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Aminopeptidase Protein
A cDNA sequence encoding the Aminopeptidase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10519-100U
Additional Details : Weight : 0.01000kg
Specifications
Aminopeptidase Protein | |
Research Use Only | |
Recombinant Aeromonas Aminopeptidase was found to have an activity of 120 Units/mg protein. | |
Other | |
Untagged | |
Buffered solution containing 10mM Tris-HCl, 100mM NaCl and 5microm ZnSO4, pH 8.0. |
100 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Non-species Specific | |
MKYTKTLLAM VLSATFCQAY AEDKVWISIG ADANQTVMKS GAESILPNSV ASSGQVWVGQ VDVAQLAELS HNMHEEHNRC GGYMVHPSAQ SAMAASAMPT TLASFVMPPI TQQATVTAWL PQVDASQITG TISSLESFTN RFYTTTSGAQ ASDWIASEWQ ALSASLPNAS VKQVSHSGYN QKSVVMTITG SEAPDEWIVI GGHLDSTIGS HTNEQSVAPG ADDDASGIAA VTEVIRVLSE NNFQPKRSIA FMAYAAEEVG LRGSQDLANQ YKSEGKNVVS ALQLDMTNYK GSAQDVVFIT DYTDSNFTQY LTQLMDEYLP SLTYGFDTCG YACSDHASWH NAGYPAAMPF ESKFNDYNPR IHTTQDTLAN SDPTGSHAKK FTQLGLAYAI EMGSATGDTP TPGNQLEDGV PVTDLSGSRG SNVWYTFELE TQKNLQITTS GGYGDLDLYV KFGSKASKQN WDCRPYLSGN NEVCTFNNAS PGTYSVMLTG YSNYSGASLK ASTF | |
Greater than 95.0% as determined by SEC-HPLC. |