missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Ag85A Protein
A cDNA sequence encoding the Ag85A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10976-10ug
Additional Details : Weight : 0.01000kg
Specifications
Ag85A Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPAFSRPGL PVEYLQVPSP SMGRDIKVQF QSGGANSPAL YLLDGLRAQD DFSGWDINTP AFEWYDQSGL SVVMPVGGQS SFYSDWYQPA CGKAGCQTYK WETFLTSELP GWLQANRHVK PTGSAVVGLS MAASSALTLA IYHPQQFVYA GAMSGLLDPS QAMGPTLIGL AMGDAGGYKA SDMWGPKEDP AWQRNDPLLN VGKLIANNTR VWVYCGNGKP SDLGGNNLPA KFLEGFVRTS NIKFQDAYNA GGGHNGVFDF PDSGTHSWEY WGAQLNAMKP DLQRALGATP NTGPAPQGAH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The Ag85A solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |