missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Multi Species PKACa2- RIa2 Protein

A cDNA sequence encoding the PKACa2- RIa2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10823-3ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955278

  • £155.00 / 3µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Multi Species PKACa2- RIa2 Protein
Research Use Only
Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
Multi Species
Untagged
Greater than 95.0% as determined by SDS-PAGE.
3 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
PKA holoenzyme type-I alpha is supplied in 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers