missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species PKACa2- RIa2 Protein
A cDNA sequence encoding the PKACa2- RIa2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10823-1ug
Additional Details : Weight : 0.01000kg
Specifications
Multi Species PKACa2- RIa2 Protein | |
Research Use Only | |
Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits. | |
Multi Species | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
PKA holoenzyme type-I alpha is supplied in 50% glycerol. |