missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Multi Species Buserelin Protein
Recombinant Protein
£99.15 - £865.00
Specifications
Name | Multi Species Buserelin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Multi Species |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15926508
|
enQuireBio™
QP11214-1MG |
1 mg |
£99.15
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946508
|
enQuireBio™
QP11214-5MG |
5 mg |
£292.00
5mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936508
|
enQuireBio™
QP11214-20MG |
20 mg |
£865.00
20mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Multi Species Buserelin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Multi Species | |
The Buserelin peptide was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |