missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Wnt-3a Protein
Click to view available options
Quantity:
10 μg
2 μg
500 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Unit Size:
10µg
2µg
500µg
Description
The Recombinant Mouse Wnt-3a Protein is derived from CHO. The Recombinant Mouse Wnt-3a Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
| Gene ID (Entrez) | 22416 |
| Molecular Weight (g/mol) | 37 kDa |
| Name | Wnt-3a |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 75%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction