missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse VEGF-164 Isoform Protein

Recombinant Protein

Brand:  enQuireBio™ QP5214-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15995699

  • £2495.00 / 500µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001273986.1
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
HUVEC proliferation was significantly induced by less than 10 ng/ml, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values of new lots are generally lower, around 5 ng/ml.
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
VEGF-164 Isoform Protein
Research Use Only
VEGFA
Recombinant Protein
E. coli
MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPENHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers