missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TSC22 domain family protein 1

A DNA sequence encoding the Mus musculus (Mouse) TSC22 domain family protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9922-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049021

  • £1930.00 / 500µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_076399.4
TSC22 domain family protein 1
Research Use Only
Tsc22d4
Recombinant Protein
Yeast
MSGGKKKSSF QITSVTTDYE GPGSPGASDS PVPPALAGPP PRLPNGDPNP DPGGRGTPRN GSPPPGAPAS RFRVVKLPQG LGEPYRRGRW TCVDVYERDL EPPSFGRLLE GIRGASGGTG GRSLDSRLEL ASLGISTPIP QPGLSQGPTS WLRPPPTSPG PQARSFTGGL GQLAGPGKAK VETPPLSASP PQQRPPGPGT GDSAQTLPSL RVEVESGGSA AATPPLSRRR DGAVRLRMEL VAPAETGKVP PTDSRPNSPA LYFDASLVHK SPDPFGAAAA QSLSLARSML AISGHLDSDD DSGSGSLVGI DNKIEQAMDL VKSHLMFAVR EEVEVLKEQI RDLAERNAAL EQENGLLRAL ASPEQLAQLP SSGLPRLGPS APNGPSI
Greater than 90% as determined by SDS-PAGE.
78829
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers