missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNFR2 Protein
A cDNA sequence encoding the TNFR2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 21938 |
---|---|
Name | TNFR2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Tnfrsf1b |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975468
|
enQuireBio™
QP10886-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965468
|
enQuireBio™
QP10886-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955468
|
enQuireBio™
QP10886-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
21938 | |
Research Use Only | |
Tnfrsf1b | |
Recombinant Protein | |
E. coli | |
MPAQVAFTPY APEPGSTCRL REYYDQTAQM CCSKCSPGQH AKVFCTKTSD TVCDSCEDST YTQLWNWVPE CLSCGSRCSS DQVETQACTR EQNRICTCRP GWYCALSKQE GCRLCAPLRK CRPGFGVARP GTETSDVVCK PCAPGTFSNT TSSTDICRPH QICNVVAIPG NASMDAVCTS TSPT | |
Greater than 97.0% as determined by SDS-PAGE. |
TNFR2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by its ability to inhibit the TNF-? mediated cytotoxicity in the L-929 cells is less than 0.2microg/ml, corresponding to a specific activity of > 5000IU/mg in the presence of 0.25ng/mL of rHuTNF-a. | |
Mouse | |
Untagged | |
TNFR2 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |