missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombopoietin / THPO Protein
Recombinant Protein
Brand: enQuireBio™ QP5439-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001166976.1 | |
Thrombopoietin / THPO Protein | |
Research Use Only | |
THPO | |
Recombinant Protein | |
E. coli | |
MSPVAPACDP RLLNKLLRDS HLLHSRLSQC PDVDPLSIPV LLPAVDFSLG EWKTQTEQSK AQDILGAVSL LLEGVMAARG QLEPSCLSSL LGQLSGQVRL LLGALQGLLG TQLPLQGRTT AHKDPNALFL SLQQLLRGKV RFLLLVEGPT LCVRRTLPTT AVPSSTSQLL TLNKF | |
Greater than 95% as determined by SDS-PAGE. |
21832 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
MO7e cell proliferation significantly induced by less than 5 ng/ml (activity of at least 2.0 x 10^5 units/mg). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |