missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Survivin / BIRC5 / API4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001012273.1 |
---|---|
Gene ID (Entrez) | 11799 |
Name | Survivin / BIRC5 / API4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967249
|
enQuireBio™
QP5720-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987249
|
enQuireBio™
QP5720-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977249
|
enQuireBio™
QP5720-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15997249
|
enQuireBio™
QP5720-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15907259
|
enQuireBio™
QP5720-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15917259
|
enQuireBio™
QP5720-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001012273.1 | |
Survivin / BIRC5 / API4 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11799 | |
Research Use Only | |
BIRC5 | |
Recombinant Protein | |
E. coli | |
MGAPALPQIW QLYLKNYRIA TFKNWPFLED CACTPERMAE AGFIHCPTEN EPDLAQCFFC FKELEGWEPD DNPIEEHRKH SPGCAFLTVK KQMEELTVSE FLKLDRQRAK NKIAKETNNK QKEFEETAKT TRQSIEQLAA | |
Greater than 90% as determined by SDS-PAGE. |