missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SHH / Sonic hedgehog Protein
Recombinant Protein
£1765.00 - £2865.00
Specifications
Accession Number | NP_033196.1 |
---|---|
Gene ID (Entrez) | 20423 |
Name | SHH / Sonic hedgehog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946179
|
enQuireBio™
QP5463-500UG |
500 μg |
£1765.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936179
|
enQuireBio™
QP5463-1MG |
1 mg |
£2865.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_033196.1 | |
SHH / Sonic hedgehog Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Sonic Hedgehog ( SHH ) activity was confirmed in a CCL-226 cell proliferatin assay. See figure for results. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
20423 | |
Research Use Only | |
SHH | |
Recombinant Protein | |
E. coli | |
MIIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKITRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRAVDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG | |
Greater than 95% as determined by SDS-PAGE. |