missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serum amyloid A-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-1 protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6648-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033143.1 | |
Serum amyloid A-1 Protein | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
E. coli | |
GFFSFVHEAF QGAGDMWRAY TDMKEANWKN SDKYFHARGN YDAAQRGPGG VWAAEKISDG REAFQEFFGR GHEDTIADQE ANRHGRSGKD PNYYRPPGLP DKY | |
Greater than 87.6% as determined by SDS-PAGE. |
20208 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |