missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse SAMHD1 Protein

A DNA sequence encoding the Mus musculus (Mouse) SAMHD1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6650-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004820

  • £545.48 / 100µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001132992.1
SAMHD1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
56045
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SAMHD1
Recombinant Protein
E. coli
DIMITDAFLK ADPYVEITGT AGKKFRISTA IDDMEAFTKL TDNIFLEVLH STDPQLSEAQ SILRNIECRN LYKYLGETQP KREKIRKEEY ERLPQEVAKA KPEKAPDVEL KAEDFIVDVI NVDYGMEDKN PIDRVHFYCK SNSKQAVRIN KEQVSQLLPE KFAEQLIRVY CKKKDGKSLD AAGKHFVQWC ALRDFTKPQD GDIIAPLITP LKWNNKTSSC LQEVSKVKTC LK
Videos
Documents