missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A5 Protein
A cDNA sequence encoding the S100A5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13395-5ug
Additional Details : Weight : 0.01000kg
Specifications
S100A5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMETPLE KALTTMVTTF HKYSGREGSK LTLSRKELKE LIKTELSLAE KMKESSIDNL MKSLDKNSDQ EIDFKEYSVF LTTLCMAYND FFLEDNK | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
S100A5 protein (1 mg/ml) is supplied in 20mM Tris-HCl buffer, pH 8.0, 40% glycerol, 3mM DTT and 200mM NaCl. |