missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ptprn2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Ptprn2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9357-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035345.2 | |
Ptprn2 Protein | |
Research Use Only | |
Ptprn2 | |
Recombinant Protein | |
Yeast | |
RGRQLPGRLG CLFEDGLCGS LETCVNDGVF GRCQKVPVMD TYRYEVPPGA LLHLKVTLQK LSRTGFTWQD DYTQRVIAQE LANLPKAYLW HGEASGPARS LQQNADNEKW FSLEREVALA KTLRRYLPYL ELLSQTPTAN AHSRIDHETR PAKGEDSSPE NILTYVAHTS ALTYPPATRA KYPDNLLRPF SRLQPDELSP KVDGDIDKQK LIAALGAYTA QRLPGENDPE PRYLVHGSAR APRPFSATAL SQRWPPPPGD AKDSPSMDDD TLLQSLLKDL QQNSEVDRLG PLKEEKADSV AGAIQSDPAE GSQESHGRGA EGQPREQTDA PETMLQDHRL SEVDDPVYKE VNRLSFQLGD LLKDYGSPLL PEGPLLEKSS REEIKKSEQP EEVLSSEEET AGVEHVRSRT YSKDLFERKP NSEPQPRRLE DQFQNRAPEL WEDEESLKLA AQGPPSGGLQ LEVQPSEEQQ GYILTGNNPL SPEKGKQLMD QVAHILRVPS SFFADIKVLG PAVTFKVSAN IQNMTTADVI KAAADNKDQL EKATGLTILQ SGIRPKGKLK LLPHQEEQED STK | |
Greater than 90% as determined by SDS-PAGE. |
19276 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |