missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRSS22 Protein
A cDNA sequence encoding the PRSS22 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13174-1mg
Additional Details : Weight : 0.01000kg
Specifications
PRSS22 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ATIRVSPDCG KPQQLNRIVG GEDSMDAQWP WIVSILKNGS HHCAGSLLTN RWVVTAAHCF KSNMDKPSLF SVLLGAWKLG SPGPRSQKVG IAWVLPHPRY SWKEGTHADI ALVRLEHSIQ FSERILPICL PDSSVRLPPK TDCWIAGWGS IQDGVPLPHP QTLQKLKVPI IDSELCKSLY WRGAGQEAIT EGMLCAGYLE GERDACLGDS GGPLMCQVDD HWLLTGIISW GEGCAERNRP GVYTSLLAHR SWVQRIVQGV QLRGYLADSG DTGSSLEHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
PRSS22 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |