missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRSS22 Protein
A cDNA sequence encoding the PRSS22 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | PRSS22 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962729
|
enQuireBio™
QP13174-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942729
|
enQuireBio™
QP13174-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952729
|
enQuireBio™
QP13174-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PRSS22 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
PRSS22 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ATIRVSPDCG KPQQLNRIVG GEDSMDAQWP WIVSILKNGS HHCAGSLLTN RWVVTAAHCF KSNMDKPSLF SVLLGAWKLG SPGPRSQKVG IAWVLPHPRY SWKEGTHADI ALVRLEHSIQ FSERILPICL PDSSVRLPPK TDCWIAGWGS IQDGVPLPHP QTLQKLKVPI IDSELCKSLY WRGAGQEAIT EGMLCAGYLE GERDACLGDS GGPLMCQVDD HWLLTGIISW GEGCAERNRP GVYTSLLAHR SWVQRIVQGV QLRGYLADSG DTGSSLEHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |