missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein FAM111A Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein FAM111A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9921-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001333474.1 | |
Protein FAM111A Protein | |
Research Use Only | |
Fam111a | |
Recombinant Protein | |
Yeast | |
MSCKKRKSQI SFNPRKNKKI KDYFSQVPKE EQNDPNTVKV DSKKMPRDIT NTRDQRPLSP RKTRQDQTPP LNKKITVTLG VNSRKHKNMK YELTCRETSS LYAALNTLSA VREEVESQKG REMLVCGKEG IEGYLNLGMP VCCIPEGSHV VITFCQCKSK TQENKQFFES QDQASTNYVR FCIHAVGSKR KKILKCGELQ KEGNKLCVYG FKGETIRDTL RKDGRFCTFI ESDDWKLIND LDTIIENTQP VDELEGKLFQ VAAELPKNPR VVSVTQNSGS ENRNFHKLEE YIVNEYTTLK EEGKKLRAYI KEKSEKRKKK ASLFKVHKEH FGKMTRNSTP VKVVKHLSRV SDSVGFLWWN NNGNAGCATC FVFKELYILT CQHVIASIVG EGIDSSEWAN IISQCVKVTF DYEELLPTGD KFFMVKPWFE ISDKHLDYAV LELKENGQEV PAGLYHRIRP VPHSGLIYII GHPEGEKKSI DCCTVVPQSS RRKKCQENFQ AREEAGFCFS TSFIHMYTQR SFQEMLHNSD VVTYDTSFFG GSSGSPVFDS NGSLVAMHAA GITCTYQAGV SNIIEFGSIM ESIDDHMKQD KYKEWYNTIS GNVQNVEMLS IDF | |
Greater than 90% as determined by SDS-PAGE. |
107373 | |
100 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |