missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein disulfide-isomerase A2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein disulfide-isomerase A2, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_001074539.1 |
---|---|
Gene ID (Entrez) | 69191 |
Name | Protein disulfide-isomerase A2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083600
|
enQuireBio™
QP6479-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023610
|
enQuireBio™
QP6479-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093600
|
enQuireBio™
QP6479-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013610
|
enQuireBio™
QP6479-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033610
|
enQuireBio™
QP6479-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003610
|
enQuireBio™
QP6479-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001074539.1 | |
Protein disulfide-isomerase A2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
69191 | |
Research Use Only | |
Pdia2 | |
Recombinant Protein | |
Yeast | |
QGEEPGGPSE VLPEEPTGEE VPKEDGILVL NHRTLSLALQ EHSALMVEFY APWCGHCKEL APEYSKAAAL LAAESAVVTL AKVDGPAEPE LTKEFEVVGY PTLKFFQNGN RTNPEEYAGP KTAEGIAEWL RRRVGPSATH LEDEEGVQAL MAKWDMVVIG FFQDLQGKDM ATFLALAKDA LDMTFGFTDQ PQLFEKFGLT KDTVVLFKKF DEGRADFPVD KETGLDLGDL SRFLVIHSMH LVTEFNSQTS PKIFAAKILN HLLLFVNQTL AQHRELLTDF REAAPPFRGQ VLFVMVDVAA DNSHVLNYFG LKAEEAPTLR LINVETTKKY APTGVIAITA ASVAAFCQAV LHGEIKHYLL SQEIPPDWDQ GPVKTLVSKN FEQVAFDETK NVFVKFYAPW CSHCKEMAPA WEALAEKYKD REDIVIAELD ATANELEAFS VLGYPTLKFF PAGPDRKVID YKSTRDLETF SKFLDSGGHL PKEEPKEPAA SAPEAQANST LGPKEEL | |
Greater than 90% as determined by SDS-PAGE. |