missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Properdin Protein

A DNA sequence encoding the Mus musculus (Mouse) Properdin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5826-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958049

  • £1930.00 / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032849.2
Properdin Protein
Research Use Only
Cfp
Recombinant Protein
Yeast
SDPVLCFTQY EESSGRCKGL LGRDIRVEDC CLNAAYAFQE HDGGLCQACR SPQWSAWSLW GPCSVTCSEG SQLRHRRCVG RGGQCSENVA PGTLEWQLQA CEDQPCCPEM GGWSEWGPWG PCSVTCSKGT QIRQRVCDNP APKCGGHCPG EAQQSQACDT QKTCPTHGAW ASWGPWSPCS GSCLGGAQEP KETRSRSCSA PAPSHQPPGK PCSGPAYEHK ACSGLPPCPV AGGWGPWSPL SPCSVTCGLG QTLEQRTCDH PAPRHGGPFC AGDATRNQMC NKAVPCPVNG EWEAWGKWSD CSRLRMSINC EGTPGQQSRS RSCGGRKFNG KPCAGKLQDI RHCYNIHNCI MKGSWSQWST WSLCTPPCSP NATRVRQRLC TPLLPKYPPT VSMVEGQGEK NVTFWGTPRP LCEALQGQKL VVEEKRSCLH VPVCKDPEEK KP
Greater than 90% as determined by SDS-PAGE.
18636
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers