missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Properdin Protein
A DNA sequence encoding the Mus musculus (Mouse) Properdin, was expressed in the hosts and tags indicated.
£261.83 - £2888.42
Specifications
Accession Number | NP_032849.2 |
---|---|
Gene ID (Entrez) | 18636 |
Name | Properdin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15918049
|
enQuireBio™
QP5826-YE-100UG |
100 μg |
£670.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958049
|
enQuireBio™
QP5826-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928049
|
enQuireBio™
QP5826-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15948049
|
enQuireBio™
QP5826-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968049
|
enQuireBio™
QP5826-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15938049
|
enQuireBio™
QP5826-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032849.2 | |
Properdin Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
18636 | |
Research Use Only | |
Cfp | |
Recombinant Protein | |
Yeast | |
SDPVLCFTQY EESSGRCKGL LGRDIRVEDC CLNAAYAFQE HDGGLCQACR SPQWSAWSLW GPCSVTCSEG SQLRHRRCVG RGGQCSENVA PGTLEWQLQA CEDQPCCPEM GGWSEWGPWG PCSVTCSKGT QIRQRVCDNP APKCGGHCPG EAQQSQACDT QKTCPTHGAW ASWGPWSPCS GSCLGGAQEP KETRSRSCSA PAPSHQPPGK PCSGPAYEHK ACSGLPPCPV AGGWGPWSPL SPCSVTCGLG QTLEQRTCDH PAPRHGGPFC AGDATRNQMC NKAVPCPVNG EWEAWGKWSD CSRLRMSINC EGTPGQQSRS RSCGGRKFNG KPCAGKLQDI RHCYNIHNCI MKGSWSQWST WSLCTPPCSP NATRVRQRLC TPLLPKYPPT VSMVEGQGEK NVTFWGTPRP LCEALQGQKL VVEEKRSCLH VPVCKDPEEK KP | |
Greater than 90% as determined by SDS-PAGE. |