missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Properdin Protein
A DNA sequence encoding the Mus musculus (Mouse) Properdin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5826-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032849.2 | |
Properdin Protein | |
Research Use Only | |
Cfp | |
Recombinant Protein | |
Yeast | |
SDPVLCFTQY EESSGRCKGL LGRDIRVEDC CLNAAYAFQE HDGGLCQACR SPQWSAWSLW GPCSVTCSEG SQLRHRRCVG RGGQCSENVA PGTLEWQLQA CEDQPCCPEM GGWSEWGPWG PCSVTCSKGT QIRQRVCDNP APKCGGHCPG EAQQSQACDT QKTCPTHGAW ASWGPWSPCS GSCLGGAQEP KETRSRSCSA PAPSHQPPGK PCSGPAYEHK ACSGLPPCPV AGGWGPWSPL SPCSVTCGLG QTLEQRTCDH PAPRHGGPFC AGDATRNQMC NKAVPCPVNG EWEAWGKWSD CSRLRMSINC EGTPGQQSRS RSCGGRKFNG KPCAGKLQDI RHCYNIHNCI MKGSWSQWST WSLCTPPCSP NATRVRQRLC TPLLPKYPPT VSMVEGQGEK NVTFWGTPRP LCEALQGQKL VVEEKRSCLH VPVCKDPEEK KP | |
Greater than 90% as determined by SDS-PAGE. |
18636 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |