missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PGAM1 Protein

A cDNA sequence encoding the PGAM1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13025-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962279

  • £151.37 / 25µg
Estimated Shipment: 17-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PGAM1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
The PGAM1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAAYKL VLIRHGESAW NLENRFSGWY DADLSPAGHE EAKRGGQALR DAGYEFDICF TSVQKRAIRT LWTVLDAIDQ MWLPVVRTWR LNERHYGGLT GLNKAETAAK HGEAQVKIWR RSYDVPPPPM EPDHPFYSNI SKDRRYADLT EDQLPSCESL KDTIARALPF WNEEIVPQIK EGKRVLIAAH GNSLRGIVKH LEGLSEEAIM ELNLPTGIPI VYELDKNLKP IKPMQFLGDE ETVRKAMEAV AAQGKVKK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers