missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nuclear respiratory factor 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nuclear respiratory factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6435-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003300

  • £654.57 / 100µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001157698.1
Nuclear respiratory factor 1 Protein
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
18181
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Nrf1
Recombinant Protein
Yeast
MEEHGVTQTE HMATIEAHAV AQQVQQVHVA TYTEHSMLSA DEDSPSSPED TSYDDSDILN STAADEVTAH LAAAGPVGMA AAAAVATGKK RKRPHVFESN PSIRKRQQTR LLRKLRATLD EYTTRVGQQA IVLCISPSKP NPVFKVFGAA PLENVVRKYK SMILEDLESA LAEHAPAPQE VNSELPPLTI DGIPVSVDKM TQAQLRAFIP EMLKYSTGRG KPGWGKESCK PIWWPEDIPW ANVRSDVRTE EQKQRVSWTQ ALRTIVKNCY KQHGREDLLY AFEDQQTQTQ ATTTHSIAHL VPSQTVVQTF SNPDGTVSLI QVGTGATVAT LADASELPTT VTVAQVNYSA VADGEVEQNW ATLQGGEMTI QTTQASEATM S-QAVASLAE AAVAASQEMQ QGATVTMALN SEAAAHAVAT LAEATLQGGG QIVLSGETAA AVGALTGVQD ANGLVQIPVS MYQTVVTSLA QGNGPVQVAM APVTTRISDS AVTMDGQAVE VVTLEQ
Videos
Documents