missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nmes1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nmes1, was expressed in the hosts and tags indicated.
£439.00 - £1230.00
Specifications
Accession Number | NP_001004174.1 |
---|---|
Gene ID (Entrez) | 433470 |
Name | Nmes1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050791
|
enQuireBio™
QP7786-MA-20UG |
20 μg |
£439.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040791
|
enQuireBio™
QP7786-MA-100UG |
100 μg |
£1230.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060791
|
enQuireBio™
QP7786-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001004174.1 | |
Nmes1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
Flag | |
Tris-based buffer, 50% glycerol |
433470 | |
Research Use Only | |
Nmes1 | |
Recombinant Protein | |
Mammal | |
MGVFQILMKN KELIPLAFFI SVAATGATSF ALYALKKTDV VIDRKRNPEP WEMVDPTQPQ KLITINQQWK PVEELQKVRR ATR | |
Greater than 90% as determined by SDS-PAGE. |