missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MYL9 Protein

A cDNA sequence encoding the MYL9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12774-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941499

  • £87.35 / 5µg

Estimated Shipment
30-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MYL9 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
MYL9 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMSSKRA KAKTTKKRPQ RATSNVFAMF DQSQIQEFKE AFNMIDQNRD GFIDKEDLHD MLASLGKNPT DEYLEGMMNE APGPINFTMF LTMFGEKLNG TDPEDVIRNA FACFDEEASG FIHEDHLREL LTTMGDRFTD EEVDEMYREA PIDKKGNFNY VEFTRILKHG AKDKDD
Videos
SDS
Documents