missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Microtubule-associated protein 1
A DNA sequence encoding the Mus musculus (Mouse) Microtubule-associated protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6335-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001297563.1 | |
Microtubule-associated protein 1 | |
Research Use Only | |
Map2 | |
Recombinant Protein | |
Yeast | |
GMEGEKLPPV PFAQTFGTNL EDRKQSTEPS IVMPSIGLSA EPPAPKEPKD WFIEMPTESK KDEWGLAAPI SPGPLTPMRE KDVLEDIPRW EGKQFDSPMP SPFHGGSFTL PLDTMKNERV SEGPRPFAPV FFQSDDKVSL QDPSALATSK ESSKDEEPLK DKADKVADVS ISEVTTLLGN VHSPVVEGYV GENISGEVKV TTDQEKKETS APSVQEPTLT ETEPQTKLDE KSTVSIEEAV AKKEESFKLR DDKTGVIQTS TEQSFSKEDQ KGQEHTIDEL KQDSFPISLE QAVTDAAMTS KTLGKVTSEP EAVSERREIQ GLFEEKTADK NKLEGAGSAT IAEVEMP | |
Greater than 90% as determined by SDS-PAGE. |
17756 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |