missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCP-2 / CCL8 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5471-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946199

  • £191.37 / 10µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_067418.1
MCP-2 / CCL8 Protein
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
THP-1 chemotaxis significantly induced by less than 100 ng/ml of Mouse MCP-2 / CCL8.
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
20307
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Ccl8
Recombinant Protein
E. coli
GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers