missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCL1 homolog Protein

A DNA sequence encoding the Mus musculus (Mouse) MCL1 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6348-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002660

  • £2410.48 / 1mg

Estimated Shipment
09-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_032588.1
MCL1 homolog Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
17210
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Mcl1
Recombinant Protein
E. coli
MFGLRRNAVI GLNLYCGGAS LGAGGGSPAG ARLVAEEAKA RREGGGEAAL LPGARVVARP PPVGAEDPDV TASAERRLHK SPGLLAVPPE EMAASAAAAI VSPEEELDGC EPEAIGKRPA VLPLLERVSE AAKSSGADGS LPSTPPPPEE EEDDLYRQSL EIISRYLREQ ATGSKDSKPL GEAGAAGRRA LETLRRVGDG VQRNHETAFQ GMLRKLDIKN EGDVKSFSRV MVHVFKDGVT NWGRIVTLIS FGAFVAKHLK SVNQESFIEP LAETITDVLV RTKRDWLVKQ RGWDGFVEFF HVQDLEGG
Videos
Documents