missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse M-CSF / CSF-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) M-CSF / CSF-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8587-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001107001.1
M-CSF / CSF-1 Protein
Research Use Only
CSF1
Recombinant Protein
E. coli
KEVSEHCSHM IGNGHLKVLQ QLIDSQMETS CQIAFEFVDQ EQLDDPVCYL KKAFFLVQDI IDETMRFKDN TPNANATERL QELSNNLNSC FTKDYEEQNK ACVRTFHETP LQLLEKIKNF FNETKNLLEK DWNIFTKNCN NSFAKCSSRD VVTKPDCNCL YPKATPSSDP ASASPHQPPA PSMAPLAGLA WDDSQRTEGS SLLPSELPLR IEDPGSAKQR PPRSTCQTLE
Greater than 90% as determined by SDS-PAGE.
12977
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers