missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IMPAD1 Protein
A cDNA sequence encoding the IMPAD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10731-2ug
Additional Details : Weight : 0.01000kg
Specifications
242291 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 5,000 pmol/min/ug and is defined as the amount of enzyme that hydrolyze Adenosine 3, 5-diphosphate per minute at pH 7.5 at 25C. | |
Mouse | |
His | |
IMPAD1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
IMPAD1 Protein | |
Research Use Only | |
Impad1 | |
Recombinant Protein | |
Baculovirus | |
ADPGRFSLFG LGSEPAAGEA EVASDGGTVD LREMLAVAVL AAERGGDEVR RVRESNVLHE KSKGKTREGA DDKMTSGDVL SNRKMFYLLK TAFPNVQINT EEHVDASDKE VIVWNRKIPE DILKEIAAPK EVPAESVTVW IDPLDATQEY TEDLRKYVTT MVCVAVNGKP VLGVIHKPFS EYTAWAMVDG GSNVKARSSY NEKTPKIIVS RSHAGMVKQV ALQTFGNQTS IIPAGGAGYK VLALLDVPDM TQEKADLYIH VTYIKKWDIC AGNAILKALG GHMTTLNGEE ISYTGSDGIE GGLLASIRMN HQALVRKLPD LEKSGHHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |