missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL36G Protein
A cDNA sequence encoding the IL36G was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10729-10ug
Additional Details : Weight : 0.01000kg
Specifications
215257 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by inducing IL-6 secretion in murine NIH/3T3 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0M 105 IU/mg. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 20mM Tris-HCl, pH 8.5, 150mM NaCl and Tween-20. |
IL36G Protein | |
Research Use Only | |
Il36g | |
Recombinant Protein | |
E. coli | |
GRETPDFGEV FDLDQQVWIF RNQALVTVPR SHRVTPVSVT ILPCKYPESL EQDKGIAIYL GIQNPDKCLF CKEVNGHPTL LLKEEKILDL YHHPEPMKPF LFYHTRTGGT STFESVAFPG HYIASSKTGN PIFLTSKKGE YYNINFNLDI KS | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |