missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL33 / Interleukin-33 / NF-HEV Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_001158196.1 |
---|---|
Gene ID (Entrez) | 77125 |
Name | IL33 / Interleukin-33 / NF-HEV Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946139
|
enQuireBio™
QP5444-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926139
|
enQuireBio™
QP5444-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916139
|
enQuireBio™
QP5444-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956139
|
enQuireBio™
QP5444-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936139
|
enQuireBio™
QP5444-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001158196.1 | |
IL33 / Interleukin-33 / NF-HEV Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2 ng/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^5 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
77125 | |
Research Use Only | |
IL33 | |
Recombinant Protein | |
E. coli | |
MSIQGTSLLT QSPASLSTYN DQSVSFVLEN GCYVINVDDS GKDQEQDQVL LRYYESPCPA SQSGDGVDGK KLMVNMSPIK DTDIWLHAND KDYSVELQRG DVSPPEQAFF VLHKKSSDFV SFECKNLPGT YIGVKDNQLA LVEEKDESCN NIMFKLSKI | |
Greater than 90 as determined by SDS-PAGE. |