missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL19 / Interleukin-19 Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_001009940.1 |
---|---|
Gene ID (Entrez) | 329244 |
Name | IL19 / Interleukin-19 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946229
|
enQuireBio™
QP5483-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926229
|
enQuireBio™
QP5483-100UG |
100 μg |
£191.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956229
|
enQuireBio™
QP5483-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936229
|
enQuireBio™
QP5483-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001009940.1 | |
IL19 / Interleukin-19 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLRRCLISVD MRLIEKSFHE IKRAMQTKDT FKNVTILSLE NLRSIKPGDV CCMTNNLLTF YRDRVFQDHQ ERSLEVLRRI SSIANSFLCV QKSLERCQVH RQCNCSQEAT NATRIIHDNY NQLEVSSAAL KSLGELNILL AWIDRNHLET PAA | |
Greater than 95% as determined by SDS-PAGE. |
329244 | |
Research Use Only | |
IL19 | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |