Learn More
enQuireBio™ Recombinant Mouse IL-31 / IL31 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5477-10ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15966209
  • £191.37 / 10µg
Estimated Shipment: 14-01-2022
Specifications
NP_083870.1
IL-31 / IL31 Protein
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse IL-31 / IL31 induces IL-8 production in human PBMCs which was detected via ELISA (typical ED50 is ? 25 ng/ml).
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
76399
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL31
Recombinant Protein
E. coli
MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C