missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-31 / IL31 Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_083870.1 |
---|---|
Gene ID (Entrez) | 76399 |
Name | IL-31 / IL31 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986209
|
enQuireBio™
QP5477-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966209
|
enQuireBio™
QP5477-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956209
|
enQuireBio™
QP5477-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996209
|
enQuireBio™
QP5477-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976209
|
enQuireBio™
QP5477-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_083870.1 | |
IL-31 / IL31 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL-31 / IL31 induces IL-8 production in human PBMCs which was detected via ELISA (typical ED50 is ? 25 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
76399 | |
Research Use Only | |
IL31 | |
Recombinant Protein | |
E. coli | |
MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C | |
Greater than 95% as determined by SDS-PAGE. |