missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-17AF Protein
Recombinant Protein
Brand: enQuireBio™ QP5205-25ug
Additional Details : Weight : 0.01000kg
Specifications
IL-17AF Protein | |
Research Use Only | |
IL17 | |
Recombinant Protein | |
E. coli | |
IL-17A:MAA IIPQSSACPN TEAKDFLQNV KVNLKVFNSL GAKVSSRRPS DYLNRSTSPW TLHRNEDPDR YPSVIWEAQC RHQRCVNAEG KLDHHMNSVL IQQEILVLKR EPESCPFTFR VEKMLVGVGC TCVASIVRQA A IL-17F:MR KNPKAGVPAL QKAGNCPPLE DNTVRVDIRI FNQNQGISVP REFQNRSSSP WDYNITRDPH RFPSEIAEAQ CRHSGCINAQ GQEDSTMNSV AIQQEILVLR REPQGCSNSF RLEKMLLKVG CTCVKPIVHQ AA | |
Greater than 95% as determined by SDS-PAGE. |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse IL-17AF bioactivity was tested by measuring IL-6 production in 3T3 cells. IL-17AF activity was measurable at concentrations above 50 ng/ml. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |