missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hmgcs1, cytoplasmic Protein
A DNA sequence encoding the Mus musculus (Mouse) Hmgcs1, cytoplasmic, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7817-ye-1mg
Additional Details : Weight : 0.01000kg
Specifica
NP_001278368.1 | |
Hmgcs1, cytoplasmic Protein | |
Research Use Only | |
Hmgcs1 | |
Recombinant Protein | |
Yeast | |
MPGSLPLNAE ACWPKDVGIV ALEIYFPSQY VDQAELEKYD GVDAGKYTIG LGQARMGFCT DREDINSLCL TVVQKLMERH SLSYDCIGRL EVGTETIIDK SKSVKSNLMQ LFEESGNTDI EGIDTTNACY GGTAAVFNAV NWVESSSWDG RYALVVAGDI AIYATGNARP TGGVGAVALL IGPNAPLIFD RGLRGTHMQH AYDFYKPDML SEYPVVDGKL SIQCYLSALD RCYSVYRKKI RAQWQKEGKD KDFTLNDFGF MIFHSPYCKL VQKSLARMFL NDFLNDQNRD KNSIYSGLEA FGDVKLEDTY FDRDVEKAFM KASSELFNQK TKASLLVSNQ NGNMYTSSVY GSLASVLAQY SPQQLAGKRV GVFSYGSGLA ATLYSLKVTQ DATPGSALDK ITASLCDLKS RLDSRTCVAP DVFAENMKLR EDTHHLANYI PQCSIDSLFE GTWYLVRVDE KHRRTYARRP FTNDHSLDEG MGLVHSNTAT EHIPSPAKKV PRLPATSAES ESAVISNGEH | |
Greater than 90% as determined by SDS-PAGE. |
208715 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |