missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GDA Protein

A cDNA sequence encoding the GDA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10627-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944458

  • £155.00 / 5µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GDA Protein
Research Use Only
Specific activity is > 4,000 pmol/min/ug, and is defined asthe amount of enzyme that convert guanine to xanthine per minute at pH 8.0 at 37C.
Mouse
His
GDA protein solution (0.5 mg/ml) containing 20mM Tris-HCl(pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol .
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMCAARTP PLALVFRGTF VHSTWTCPME VLRDHLLGVS DSGKIVFLEE SSQQEKLAKE WCFKPCEIRE LSHHEFFMPG LVDTHIHAPQ YAFAGSNVDL PLLEWLNKYT FPTEQRFRST DVAEEVYTRV VRRTLKNGTT TACYFGTIHT DSSLILAEIT DKFGQRAFVG KVCMDLNDTV PEYKETTEES VKETERFVSE MLQKNYPRVK PIVTPRFTLS CTETLMSELG NIAKTHDLYI QSHISENREE IEAVKSLYPS YKNYTDVYDK NNLLTNKTVM AHGCYLSEEE LNIFSERGAS IAHCPNSNLS LSSGLLNVLE VLKHKVKIGL GTDVAGGYSY SMLDAIRRAV MVSNVLLINK VNEKNLTLKE VFRLATLGGS QALGLDSEIG NFEVGKEFDA LLINPRASDS PIDLFYGDFV GDISEAVIQK FLYLGDDRNI EEVYVGGKQV VPFSSSV
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers