missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse G-CSF / CSF3 Protein
Recombinant Protein
£605.00 - £3965.00
Specifications
Accession Number | NP_034101.1 |
---|---|
Gene ID (Entrez) | 12985 |
Name | G-CSF / CSF3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946089
|
enQuireBio™
QP5415-100UG |
100 μg |
£605.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966089
|
enQuireBio™
QP5415-500UG |
500 μg |
£2425.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956089
|
enQuireBio™
QP5415-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034101.1 | |
G-CSF / CSF3 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 50 pg/mL in the NFS-60 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^7 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium citrate, pH 3.0 |
12985 | |
Research Use Only | |
CSF3 | |
Recombinant Protein | |
E. coli | |
MVPLVTVSAL PPSLPLPRSF LLKSLEQVRK IQASGSVLLE QLCATYKLCH PEELVLLGHS LGIPKASLSG CSSQALQQTQ CLSQLHSGLC LYQGLLQALS GISPALAPTL DLLQLDVANF ATTIWQQMEN LGVAPTVQPT QSAMPAFTSA FQRRAGGVLA ISYLQGFLET ARLALHHLA | |
Greater than 95% as determined by SDS-PAGE. |