missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ESM1 / Endocan Protein
A DNA sequence encoding the Mus musculus (Mouse) ESM1 / Endocan, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5997-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_076101.1 | |
ESM1 / Endocan Protein | |
Research Use Only | |
ESM1 | |
Recombinant Protein | |
E. coli | |
AKYAVDCPEH CDKTECRSSL RCKRTVLDDC GCCQVCAAGP GETCYRTVSG MDGVKCGPGL KCHFYSEEDD FGDEFGICKD CPYGTFGMEC KETCNCQSGI CDRVTGRCLD FPFFQYAAAK SPSRTSASHT ERDSASGDGN AVREEIGEGN AARPSVMKWL NPR | |
Greater than 90% as determined by SDS-PAGE. |
71690 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |