missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ErbB3 Protein

A cDNA sequence encoding the ErbB3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11804-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968428

  • £5730.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ErbB3 Protein
Research Use Only
Recombinant Protein
Baculovirus
SEMGNSQAVC PGTLNGLSVT GDADNQYQTL YKLYEKCEVV MGNLEIVLTG HNADLSFLQW IREVTGYVLV AMNEFSVLPL PNLRVVRGTQ VYDGKFAIFV MLNYNTNSSH ALRQLRFTQL TEILLGGVYI EKNDKLCHMD TIDWRDIVRV PDAEIVVKNN GGNCPPCHEV CKGRCWGPGP EDCQILTKTI CAPQCNGRCF GPNPNQCCHD ECAGGCSGPQ DTDCFACRHF NDSGACVPRC PAPLVYNKLT FQLEPNPHIK YQYGGVCVAS CPHNFVVDQT FCVRACPADK MEVDKNGLKM CEPCRGLCPK ACEGTGSGSR YQTVDSSNID GFVNCTKILG NLDFLITGLN GDPWHKIPAL DPEKLNVFRT VREITGYLNI QSWPPHMHNF SVFSNLTTIG GRSLYNRGFS LLIMKNLNVT SLGFRSLKEI SAGRVYISAN QQLCYHHSLN WTRLLRGPAE ERLDIKYNRP LGECVAEGKV CDPLCSSGGC WGPGPGQCLS CRNYSREGVC VTHCNVLQGE PREFVHEAHC FSCHPECQPM EGTSTCNGSG SDACARCAHF RDGPHCVNSC PHGILGAKGP IYKYPDAQNE CRPCHENCTQ GCKGPELQDC LGQAEVLMSK PHLEHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
ErbB3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers