missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Endoplasmic reticulum aminopeptidase 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Endoplasmic reticulum aminopeptidase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5992-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939199

  • £560.00 / 100µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_109636.1
Endoplasmic reticulum aminopeptidase 1 Protein
Research Use Only
Erap1
Recombinant Protein
E. coli
PCVQRAERYF REWKSSNGNM SIPIDVTLAV FAVGAQNTEG WDFLYSKYQS SLSSTEKSQI EFSLCTSKDP EKLQWLLDQS FKGEIIKTQE FPHILTLIGR NPVGYPLAWK FLRENWNKLV QKFELGSSSI AHMVMGTTDQ FSTRARLEEV KGFFSSLKEN GSQLRCVQQT IETIEENIRW MDKNFDKIRL WLQKEKPELL
Greater than 80.6% as determined by SDS-PAGE.
80898
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers