missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EFNB2 Protein
A cDNA sequence encoding the EFNB2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11755-50ug
Additional Details : Weight : 0.01000kg
Specifications
EFNB2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
RSIVLEPIYW NSSNSKFLPG QGLVLYPQIG DKLDIICPKV DSKTVGQYEY YKVYMVDKDQ ADRCTIKKEN TPLLNCARPD QDVKFTIKFQ EFSPNLWGLE FQKNKDYYII STSNGSLEGL DNQEGGVCQT RAMKILMKVG QDASSAGSAR NHGPTRRPEL EAGTNGRSST TSPFVKPNPG SSTDGNSAGH SGNNLLGSEV ALFALEHHHH HH | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The EFNB2 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |