missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Desmoglein-2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Desmoglein-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5947-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_031909.2 | |
Desmoglein-2 Protein | |
Research Use Only | |
Dsg2 | |
Recombinant Protein | |
E. coli | |
AWITAPVALR EGEDLSRKNP IAKIHSDLAE EKGIKITYKY TGKGITEPPF GIFVFDRNTG ELNITSILDR EETPYFLLTG YALDSRGNNL EKPLELRIKV LDINDNEPVF TQEVFVGSIE ELSAAHTLVM KITATDADDP ETLNAKVSYR IVSQEPANSH MFYLNKDTGE IYTTSFTLDR EEHSSYSLTV EARDGNGQIT DKPVQQAQVQ IRILDVNDNI PVVENKMYEG TVEENQVNVE VMRIKVTDAD EVGSDNWLAN FTFASGNEGG YFHIETDTQT NEGIVTLVKE VDYEEMKKLD LSIIVTNKAA FHKSILSKYK ATPIPITVKV KNVVEGIHFK SSVVSFRASE AMDRSSLSRS IGNFQVFDED TGQAAKVTYV KVQDTDNWVS VDSVTSEIKL VKIPDFESRY VQNGTYTAKV VAISKEHPQK TITGTIVITV EDVNDNCPVL VDSVRSVCED EPYVNVTAED LDGAQNSAPF SFSIIDQPPG TAQKWKITHQ ESTSVLLQQS ERKRGRSEIP FLISDSQGFS CPERQVLQLT VCECLKGGGC VAAQYDNYVG LGPA | |
Greater than 90% as determined by SDS-PAGE. |
13511 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |