missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Desmoglein-2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Desmoglein-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5947-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15908879

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_031909.2
Desmoglein-2 Protein
Research Use Only
Dsg2
Recombinant Protein
E. coli
AWITAPVALR EGEDLSRKNP IAKIHSDLAE EKGIKITYKY TGKGITEPPF GIFVFDRNTG ELNITSILDR EETPYFLLTG YALDSRGNNL EKPLELRIKV LDINDNEPVF TQEVFVGSIE ELSAAHTLVM KITATDADDP ETLNAKVSYR IVSQEPANSH MFYLNKDTGE IYTTSFTLDR EEHSSYSLTV EARDGNGQIT DKPVQQAQVQ IRILDVNDNI PVVENKMYEG TVEENQVNVE VMRIKVTDAD EVGSDNWLAN FTFASGNEGG YFHIETDTQT NEGIVTLVKE VDYEEMKKLD LSIIVTNKAA FHKSILSKYK ATPIPITVKV KNVVEGIHFK SSVVSFRASE AMDRSSLSRS IGNFQVFDED TGQAAKVTYV KVQDTDNWVS VDSVTSEIKL VKIPDFESRY VQNGTYTAKV VAISKEHPQK TITGTIVITV EDVNDNCPVL VDSVRSVCED EPYVNVTAED LDGAQNSAPF SFSIIDQPPG TAQKWKITHQ ESTSVLLQQS ERKRGRSEIP FLISDSQGFS CPERQVLQLT VCECLKGGGC VAAQYDNYVG LGPA
Greater than 90% as determined by SDS-PAGE.
13511
500 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers