missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5454-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946159

  • £3085.00 / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_038683.1
CXCL12 / SDF-1 Protein
Research Use Only
CXCL12
Recombinant Protein
E. coli
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
Greater than 95% as determined by SDS-PAGE.
20315
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse CXCL12 / SDF-1 induces Primary human T cell chemotaxis (significanty different from control at Mouse CXCL12 / SDF-1 concentrations greater than 5-50 ng/ml).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers