missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CST3 Protein
A cDNA sequence encoding the CST3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10560-HIS-INSECT-2ug
Additional Details : Weight : 0.01000kg
Specifica
13010 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
ATPKQGPRML GAPEEADANE EGVRRALDFA VSEYNKGSND AYHSRAIQVV RARKQLVAGV NYFLDVEMGR TTCTKSQTNL TDCPFHDQPH LMRKALCSFQ IYSVPWKGTH SLTKFSCKNA HHHHHH | |
Greater than 95% as determined by SDS-PAGE. |
CST3 Protein | |
Research Use Only | |
Cst3 | |
Mouse | |
His | |
CST3 protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |